The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013302.3 | NP_037434.1 | 12 - 13 | 1862 | 91 |
XM_017023193.1 | XP_016878682.1 | 12 - 13 | 1573 | 91 | |
GenBank mRNA | AK308757.1 | - | 12 - 13 | 1939 | 91 |
BC032665.1 | - | 13 - 14 | 1573 | 91 | |
U93850.1 | - | 11 - 12 | 1377 | 91 |