The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005272.3 | NP_005263.1 | 2 - 3 | 379 | 78 |
XM_011541264.2 | XP_011539566.1 | 3 - 4 | 470 | 78 | |
GenBank mRNA | AF493909.1 | - | 2 - 3 | 166 | 78 |
BC000233.1 | - | 2 - 3 | 347 | 78 | |
BT009844.1 | - | 2 - 3 | 166 | 78 |