The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024867.3 | NP_079143.3 | 16 - 17 | 2506 | 89 |
XM_005248376.3 | XP_005248433.1 | 16 - 17 | 2611 | 89 | |
XM_005248377.3 | XP_005248434.1 | 16 - 17 | 2596 | 89 | |
XM_005248378.3 | XP_005248435.1 | 16 - 17 | 2600 | 89 | |
XM_011514135.2 | XP_011512437.1 | 16 - 17 | 2616 | 89 | |
XM_011514136.2 | XP_011512438.1 | 16 - 17 | 2615 | 89 | |
XM_011514137.2 | XP_011512439.1 | 16 - 17 | 2600 | 89 | |
XM_011514138.2 | XP_011512440.1 | 16 - 17 | 2616 | 89 | |
XM_011514139.2 | XP_011512441.1 | 16 - 17 | 2615 | 89 | |
XM_011514140.2 | XP_011512442.1 | 15 - 16 | 2350 | 89 | |
XM_011514141.2 | XP_011512443.1 | 17 - 18 | 2814 | 89 | |
XM_011514142.1 | XP_011512444.1 | 16 - 17 | 2611 | 89 | |
XM_017009880.1 | XP_016865369.1 | 15 - 16 | 2432 | 89 | |
XM_017009881.1 | XP_016865370.1 | 16 - 17 | 3470 | 89 | |
XM_017009882.1 | XP_016865371.1 | 16 - 17 | 2604 | 89 | |
GenBank mRNA | AB051557.1 | - | 6 - 7 | 847 | 89 |
AK094564.1 | - | 5 - 6 | 529 | 89 | |
AK097547.1 | - | 7 - 8 | 1247 | 89 | |
AL832607.1 | - | 8 - 9 | 4117 | 89 | |
DA295300.1 | - | 1 - 2 | 125 | 89 | |
EF553528.1 | - | 16 - 17 | 2441 | 89 |