The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024867.3 | NP_079143.3 | 20 - 21 | 3029 | 109 |
XM_005248376.3 | XP_005248433.1 | 20 - 21 | 3134 | 109 | |
XM_005248377.3 | XP_005248434.1 | 20 - 21 | 3119 | 109 | |
XM_011514135.2 | XP_011512437.1 | 20 - 21 | 3139 | 109 | |
XM_011514136.2 | XP_011512438.1 | 20 - 21 | 3138 | 109 | |
XM_011514137.2 | XP_011512439.1 | 20 - 21 | 3123 | 109 | |
XM_011514140.2 | XP_011512442.1 | 19 - 20 | 2873 | 109 | |
XM_011514141.2 | XP_011512443.1 | 21 - 22 | 3337 | 109 | |
XM_017009880.1 | XP_016865369.1 | 19 - 20 | 2955 | 109 | |
XM_017009881.1 | XP_016865370.1 | 20 - 21 | 3993 | 109 | |
XM_017009882.1 | XP_016865371.1 | 20 - 21 | 3127 | 109 | |
GenBank mRNA | AB051557.1 | - | 10 - 11 | 1370 | 109 |