Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPEF2
Entrez Gene ID
79925
Gene Name
sperm flagellar 2
Gene Aliases
CT122, KPL2
Chromosome Location
Chr.5: 35617773 - 35814611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024867.3 NP_079143.3 1 - 2 171 75
NM_144722.3 NP_653323.1 1 - 2 171 75
XM_005248376.3 XP_005248433.1 1 - 2 276 75
XM_005248377.3 XP_005248434.1 1 - 2 276 75
XM_005248378.3 XP_005248435.1 1 - 2 265 75
XM_011514135.2 XP_011512437.1 1 - 2 281 75
XM_011514136.2 XP_011512438.1 1 - 2 280 75
XM_011514137.2 XP_011512439.1 1 - 2 280 75
XM_011514138.2 XP_011512440.1 1 - 2 281 75
XM_011514139.2 XP_011512441.1 1 - 2 280 75
XM_011514140.2 XP_011512442.1 1 - 2 186 75
XM_011514141.2 XP_011512443.1 1 - 2 284 75
XM_011514142.1 XP_011512444.1 1 - 2 276 75
XM_017009880.1 XP_016865369.1 1 - 2 283 75
XM_017009881.1 XP_016865370.1 1 - 2 285 75
XM_017009882.1 XP_016865371.1 1 - 2 269 75
GenBank mRNA AK058124.1 - 1 - 2 96 75
BC110984.1 - 1 - 2 86 75
EF553528.1 - 1 - 2 124 75

View Full Product Details