Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPEF2
Entrez Gene ID
79925
Gene Name
sperm flagellar 2
Gene Aliases
CT122, KPL2
Chromosome Location
Chr.5: 35617773 - 35814611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024867.3 NP_079143.3 21 - 22 3176 124
XM_005248376.3 XP_005248433.1 21 - 22 3281 124
XM_005248377.3 XP_005248434.1 21 - 22 3266 124
XM_011514135.2 XP_011512437.1 21 - 22 3286 124
XM_011514136.2 XP_011512438.1 21 - 22 3285 124
XM_011514137.2 XP_011512439.1 21 - 22 3270 124
XM_011514138.2 XP_011512440.1 21 - 22 3262 124
XM_011514139.2 XP_011512441.1 20 - 21 3210 124
XM_011514140.2 XP_011512442.1 20 - 21 3020 124
XM_011514141.2 XP_011512443.1 22 - 23 3484 124
XM_011514142.1 XP_011512444.1 21 - 22 3257 124
XM_017009880.1 XP_016865369.1 20 - 21 3102 124
XM_017009881.1 XP_016865370.1 21 - 22 4140 124
XM_017009882.1 XP_016865371.1 21 - 22 3274 124
GenBank mRNA AB051557.1 - 11 - 12 1517 124

View Full Product Details