Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPEF2
Entrez Gene ID
79925
Gene Name
sperm flagellar 2
Gene Aliases
CT122, KPL2
Chromosome Location
Chr.5: 35617773 - 35814611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024867.3 NP_079143.3 22 - 23 3305 82
XM_005248376.3 XP_005248433.1 22 - 23 3410 82
XM_005248377.3 XP_005248434.1 22 - 23 3395 82
XM_011514135.2 XP_011512437.1 22 - 23 3415 82
XM_011514136.2 XP_011512438.1 22 - 23 3414 82
XM_011514137.2 XP_011512439.1 22 - 23 3399 82
XM_011514138.2 XP_011512440.1 22 - 23 3391 82
XM_011514139.2 XP_011512441.1 21 - 22 3339 82
XM_011514140.2 XP_011512442.1 21 - 22 3149 82
XM_011514141.2 XP_011512443.1 23 - 24 3613 82
XM_011514142.1 XP_011512444.1 22 - 23 3386 82
XM_017009880.1 XP_016865369.1 21 - 22 3231 82
XM_017009881.1 XP_016865370.1 22 - 23 4269 82
XM_017009882.1 XP_016865371.1 22 - 23 3403 82
GenBank mRNA AB051557.1 - 12 - 13 1646 82

View Full Product Details