Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPEF2
Entrez Gene ID
79925
Gene Name
sperm flagellar 2
Gene Aliases
CT122, KPL2
Chromosome Location
Chr.5: 35617773 - 35814611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024867.3 NP_079143.3 36 - 37 5487 107
XM_011514136.2 XP_011512438.1 41 - 42 6418 107
XM_011514142.1 XP_011512444.1 36 - 37 5568 107
GenBank mRNA AK026817.1 - 12 - 13 1991 107
AK027230.1 - 13 - 14 1933 107
AK225586.1 - 13 - 14 1933 107
AK225607.1 - 13 - 14 1933 107
BX648691.1 - 14 - 15 2995 107

View Full Product Details