The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016616.4 | NP_057700.3 | 16 - 17 | 1914 | 149 |
GenBank mRNA | AF202051.2 | - | 15 - 16 | 1643 | 149 |
AF305596.1 | - | 15 - 16 | 1612 | 149 | |
AK313599.1 | - | 15 - 16 | 1654 | 149 | |
BC036816.1 | - | 16 - 17 | 1914 | 149 | |
GQ472221.1 | - | 16 - 17 | 1914 | 149 |