Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
Chromosome Location
Chr.7: 37848597 - 37900401 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016616.4 NP_057700.3 3 - 4 402 99
GenBank mRNA AF202051.2 - 2 - 3 131 99
AF305596.1 - 2 - 3 100 99
BC036816.1 - 3 - 4 402 99
GQ472221.1 - 3 - 4 402 99

View Full Product Details