The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278273.1 | NP_001265202.1 | 12 - 13 | 1694 | 89 |
NM_182691.2 | NP_872633.1 | 10 - 11 | 1747 | 89 | |
NM_182692.2 | NP_872634.1 | 11 - 12 | 1670 | 89 | |
XM_005250549.3 | XP_005250606.2 | 12 - 13 | 1929 | 89 | |
XM_005250550.3 | XP_005250607.1 | 11 - 12 | 1674 | 89 | |
XM_005250551.4 | XP_005250608.1 | 12 - 13 | 1722 | 89 | |
XM_006716098.2 | XP_006716161.1 | 12 - 13 | 1689 | 89 | |
XM_011516536.2 | XP_011514838.1 | 10 - 11 | 1685 | 89 | |
XM_011516538.2 | XP_011514840.1 | 11 - 12 | 1558 | 89 | |
XM_017012560.1 | XP_016868049.1 | 12 - 13 | 1744 | 89 | |
XM_017012561.1 | XP_016868050.1 | 11 - 12 | 1594 | 89 | |
XM_017012562.1 | XP_016868051.1 | 12 - 13 | 1928 | 89 | |
XM_017012563.1 | XP_016868052.1 | 12 - 13 | 1929 | 89 | |
XM_017012564.1 | XP_016868053.1 | 10 - 11 | 1733 | 89 | |
XM_017012565.1 | XP_016868054.1 | 12 - 13 | 1693 | 89 | |
XM_017012566.1 | XP_016868055.1 | 12 - 13 | 1730 | 89 | |
GenBank mRNA | AK093420.1 | - | 2 - 3 | 1469 | 89 |
AY354201.1 | - | 11 - 12 | 1735 | 89 | |
BC035214.1 | - | 10 - 11 | 1666 | 89 | |
BC068547.1 | - | 12 - 13 | 1694 | 89 |