The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278273.1 | NP_001265202.1 | 6 - 7 | 578 | 79 |
NM_182691.2 | NP_872633.1 | 4 - 5 | 631 | 79 | |
NM_182692.2 | NP_872634.1 | 5 - 6 | 554 | 79 | |
XM_005250549.3 | XP_005250606.2 | 6 - 7 | 813 | 79 | |
XM_005250550.3 | XP_005250607.1 | 5 - 6 | 558 | 79 | |
XM_005250551.4 | XP_005250608.1 | 6 - 7 | 606 | 79 | |
XM_006716098.2 | XP_006716161.1 | 6 - 7 | 573 | 79 | |
XM_011516536.2 | XP_011514838.1 | 4 - 5 | 569 | 79 | |
XM_011516538.2 | XP_011514840.1 | 5 - 6 | 442 | 79 | |
XM_017012560.1 | XP_016868049.1 | 6 - 7 | 628 | 79 | |
XM_017012561.1 | XP_016868050.1 | 5 - 6 | 478 | 79 | |
XM_017012562.1 | XP_016868051.1 | 6 - 7 | 812 | 79 | |
XM_017012563.1 | XP_016868052.1 | 6 - 7 | 813 | 79 | |
XM_017012564.1 | XP_016868053.1 | 4 - 5 | 617 | 79 | |
XM_017012565.1 | XP_016868054.1 | 6 - 7 | 577 | 79 | |
XM_017012566.1 | XP_016868055.1 | 6 - 7 | 614 | 79 | |
GenBank mRNA | AY354201.1 | - | 5 - 6 | 619 | 79 |
BC035214.1 | - | 4 - 5 | 550 | 79 | |
BC068547.1 | - | 6 - 7 | 578 | 79 | |
BM805927.1 | - | - | 605 | 79 | |
U88666.1 | - | 4 - 5 | 584 | 79 |