Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACBD7
Entrez Gene ID
414149
Gene Name
acyl-CoA binding domain containing 7
Gene Aliases
bA455B2.2
Chromosome Location
Chr.10: 15075475 - 15088776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039844.2 NP_001034933.1 1 - 2 62 75
GenBank mRNA AK095538.1 - 1 - 2 62 75
AK295169.1 - 1 - 2 62 75
BC029526.1 - 1 - 2 62 75

View Full Product Details