Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMP3
Entrez Gene ID
27074
Gene Name
lysosomal associated membrane protein 3
Gene Aliases
CD208, DC LAMP, DC-LAMP, DCLAMP, LAMP, LAMP-3, TSC403
Chromosome Location
Chr.3: 183122213 - 183162879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014398.3 NP_055213.2 1 - 2 278 58
XM_005247360.4 XP_005247417.1 1 - 2 114 58
XM_011512688.1 XP_011510990.1 1 - 2 114 58
GenBank mRNA AB013924.1 - 1 - 2 117 58
AJ005766.1 - 1 - 2 96 58
AK313172.1 - 1 - 2 133 58
BC032940.2 - 1 - 2 234 58
DA994656.1 - 1 - 2 115 58
DC399447.1 - 1 - 2 310 58

View Full Product Details