The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015447.3 | NP_056262.3 | 13 - 14 | 4173 | 64 |
XM_005263396.3 | XP_005263453.1 | 14 - 15 | 4598 | 64 | |
XM_005263397.1 | XP_005263454.1 | 11 - 12 | 3699 | 64 | |
XM_011518272.1 | XP_011516574.1 | 13 - 14 | 3867 | 64 | |
XM_011518273.2 | XP_011516575.1 | 12 - 13 | 3698 | 64 | |
XM_011518274.1 | XP_011516576.1 | 11 - 12 | 3728 | 64 | |
XM_017014301.1 | XP_016869790.1 | 13 - 14 | 4454 | 64 | |
XM_017014302.1 | XP_016869791.1 | 13 - 14 | 4378 | 64 | |
GenBank mRNA | AJ519841.1 | - | 11 - 12 | 3698 | 64 |
AL834528.1 | - | 3 - 4 | 525 | 64 | |
BC010646.1 | - | 3 - 4 | 899 | 64 | |
BC012778.2 | - | 3 - 4 | 2173 | 64 | |
BC130580.1 | - | 13 - 14 | 3832 | 64 | |
BC144082.1 | - | 13 - 14 | 3832 | 64 |