The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015447.3 | NP_056262.3 | 2 - 3 | 429 | 55 |
XM_005263396.3 | XP_005263453.1 | 2 - 3 | 821 | 55 | |
XM_005263397.1 | XP_005263454.1 | 1 - 2 | 97 | 55 | |
GenBank mRNA | AF116701.2 | - | 2 - 3 | 408 | 55 |
AJ519841.1 | - | 1 - 2 | 96 | 55 | |
BC130580.1 | - | 1 - 2 | 55 | 55 | |
BC144082.1 | - | 1 - 2 | 55 | 55 |