The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015447.3 | NP_056262.3 | 3 - 4 | 582 | 116 |
GenBank mRNA | AF116701.2 | - | 3 - 4 | 561 | 116 |
BU949503.1 | - | 1 - 2 | 144 | 116 |