Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIMS1
Entrez Gene ID
22999
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location
Chr.6: 71886703 - 72403143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014989.5 NP_055804.2 1 - 2 482 70
XM_005248688.3 XP_005248745.1 1 - 2 418 70
XM_011535604.2 XP_011533906.2 1 - 2 418 70
XM_011535611.2 XP_011533913.2 1 - 2 418 70
XM_011535615.2 XP_011533917.2 1 - 2 418 70
XM_017010516.1 XP_016866005.1 1 - 2 418 70
XM_017010517.1 XP_016866006.1 1 - 2 418 70
XM_017010518.1 XP_016866007.1 1 - 2 418 70
XM_017010519.1 XP_016866008.1 1 - 2 418 70
XM_017010520.1 XP_016866009.1 1 - 2 418 70
XM_017010521.1 XP_016866010.1 1 - 2 418 70
XM_017010522.1 XP_016866011.1 1 - 2 418 70
XM_017010523.1 XP_016866012.1 1 - 2 418 70
XM_017010524.1 XP_016866013.1 1 - 2 418 70
XM_017010525.1 XP_016866014.1 1 - 2 418 70
XM_017010526.1 XP_016866015.1 1 - 2 418 70
XM_017010527.1 XP_016866016.1 1 - 2 418 70
XM_017010528.1 XP_016866017.1 1 - 2 418 70
XM_017010529.1 XP_016866018.1 1 - 2 418 70
XM_017010530.1 XP_016866019.1 1 - 2 418 70
XM_017010531.1 XP_016866020.1 1 - 2 418 70
XM_017010532.1 XP_016866021.1 1 - 2 418 70
XM_017010533.1 XP_016866022.1 1 - 2 418 70
GenBank mRNA AB002338.1 - 1 - 2 238 70
AB045726.1 - 1 - 2 481 70
AB051866.1 - 1 - 2 481 70
AY190519.1 - 1 - 2 161 70
BC151853.1 - 1 - 2 262 70
BC152435.1 - 1 - 2 238 70

View Full Product Details