Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIMS1
Entrez Gene ID
22999
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location
Chr.6: 71886703 - 72403143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168407.1 NP_001161879.1 14 - 15 1854 64
NM_001168408.1 NP_001161880.1 14 - 15 1857 64
NM_001168409.1 NP_001161881.1 14 - 15 1642 64
NM_001168410.1 NP_001161882.1 14 - 15 1929 64
XM_005248688.3 XP_005248745.1 19 - 20 3380 64
XM_011535604.2 XP_011533906.2 19 - 20 3377 64
XM_011535615.2 XP_011533917.2 19 - 20 3377 64
XM_017010517.1 XP_016866006.1 19 - 20 3380 64
XM_017010518.1 XP_016866007.1 19 - 20 3377 64
XM_017010520.1 XP_016866009.1 19 - 20 3380 64
XM_017010522.1 XP_016866011.1 19 - 20 3380 64
XM_017010523.1 XP_016866012.1 19 - 20 3380 64
XM_017010524.1 XP_016866013.1 19 - 20 3377 64
XM_017010527.1 XP_016866016.1 19 - 20 3377 64
XM_017010530.1 XP_016866019.1 19 - 20 3380 64
XM_017010531.1 XP_016866020.1 19 - 20 3377 64
XM_017010532.1 XP_016866021.1 19 - 20 3380 64
XM_017010534.1 XP_016866023.1 14 - 15 1953 64
XM_017010535.1 XP_016866024.1 14 - 15 1950 64
XM_017010537.1 XP_016866026.1 14 - 15 1950 64
XM_017010538.1 XP_016866027.1 14 - 15 2058 64
XM_017010539.1 XP_016866028.1 14 - 15 1945 64
XM_017010540.1 XP_016866029.1 14 - 15 1944 64
XM_017010541.1 XP_016866030.1 14 - 15 1941 64
XM_017010542.1 XP_016866031.1 14 - 15 1944 64
XM_017010543.1 XP_016866032.1 14 - 15 1860 64
XM_017010546.1 XP_016866035.1 14 - 15 2056 64
XM_017010547.1 XP_016866036.1 14 - 15 2053 64
XM_017010550.1 XP_016866039.1 14 - 15 2324 64
XM_017010551.1 XP_016866040.1 14 - 15 2321 64
XM_017010552.1 XP_016866041.1 14 - 15 2181 64
XM_017010553.1 XP_016866042.1 14 - 15 2178 64
XM_017010554.1 XP_016866043.1 14 - 15 2174 64
XM_017010557.1 XP_016866046.1 2 - 3 180 64
XM_017010558.1 XP_016866047.1 1 - 2 73 64
GenBank mRNA AB051866.1 - 20 - 21 3410 64
AF263306.1 - 7 - 8 798 64
AF263308.1 - 7 - 8 798 64
AF263309.1 - 7 - 8 795 64
AK293828.1 - 14 - 15 1532 64
AK294868.1 - 14 - 15 1854 64
AK295001.1 - 14 - 15 1775 64
AK296303.1 - 14 - 15 1698 64
AK308658.1 - 14 - 15 1929 64
AK309185.1 - 14 - 15 1642 64
AK316108.1 - 14 - 15 1655 64

View Full Product Details