The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198489.2 | NP_940891.1 | 7 - 8 | 747 | 86 |
NR_104049.1 | 7 - 8 | 775 | 86 | ||
NR_104050.1 | 7 - 8 | 775 | 86 | ||
XM_011542799.1 | XP_011541101.1 | 7 - 8 | 775 | 86 | |
XM_011542800.2 | XP_011541102.1 | 5 - 6 | 455 | 86 | |
XM_017017652.1 | XP_016873141.1 | 7 - 8 | 3529 | 86 | |
GenBank mRNA | AB094093.1 | - | 7 - 8 | 738 | 86 |
BC126469.1 | - | 7 - 8 | 712 | 86 | |
BC126471.1 | - | 7 - 8 | 712 | 86 | |
BC144209.1 | - | 7 - 8 | 740 | 86 | |
BC144210.1 | - | 7 - 8 | 740 | 86 |