The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152905.3 | NP_690869.1 | 2 - 3 | 329 | 68 |
GenBank mRNA | AK093221.1 | - | 2 - 3 | 267 | 68 |
AK290058.1 | - | 2 - 3 | 329 | 68 | |
AK315821.1 | - | 2 - 3 | 295 | 68 | |
DB074571.1 | - | 2 - 3 | 329 | 68 |