The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006724.3 | NP_006715.2 | 16 - 17 | 3728 | 79 |
XM_005266989.2 | XP_005267046.1 | 16 - 17 | 3920 | 79 | |
GenBank mRNA | AF116604.1 | - | 5 - 6 | 544 | 79 |
BC146770.1 | - | 16 - 17 | 3728 | 79 | |
D86968.2 | - | 16 - 17 | 3728 | 79 |