The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014845.5 | NP_055660.1 | 15 - 16 | 1968 | 103 |
XM_011536281.2 | XP_011534583.1 | 15 - 16 | 2500 | 103 | |
XM_017011591.1 | XP_016867080.1 | 15 - 16 | 1973 | 103 | |
XM_017011592.1 | XP_016867081.1 | 14 - 15 | 1736 | 103 | |
XM_017011593.1 | XP_016867082.1 | 13 - 14 | 1671 | 103 | |
GenBank mRNA | AK291671.1 | - | 15 - 16 | 1893 | 103 |
AK293422.1 | - | 10 - 11 | 1219 | 103 | |
BC041338.2 | - | 15 - 16 | 1924 | 103 | |
D87464.1 | - | 15 - 16 | 1877 | 103 |