The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000606.2 | NP_000597.2 | 2 - 3 | 350 | 79 |
GenBank mRNA | BC113624.1 | - | 2 - 3 | 325 | 79 |
BC113626.1 | - | 2 - 3 | 325 | 79 | |
BM852158.1 | - | 2 - 3 | 350 | 79 | |
M17263.1 | - | 2 - 3 | 301 | 79 | |
M17999.1 | - | 2 - 3 | 336 | 79 | |
X06465.1 | - | 2 - 3 | 336 | 79 |