The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006895.2 | NP_008826.1 | 3 - 4 | 559 | 75 |
XM_011511064.2 | XP_011509366.1 | 2 - 3 | 322 | 75 | |
XM_017003948.1 | XP_016859437.1 | 4 - 5 | 1019 | 75 | |
XM_017003949.1 | XP_016859438.1 | 3 - 4 | 392 | 75 | |
GenBank mRNA | AF523358.1 | - | 3 - 4 | 486 | 75 |
AF523359.1 | - | 3 - 4 | 486 | 75 | |
AF523360.1 | - | 3 - 4 | 486 | 75 | |
AK301462.1 | - | 3 - 4 | 317 | 75 | |
AK313804.1 | - | 3 - 4 | 317 | 75 | |
BC020677.1 | - | 3 - 4 | 318 | 75 | |
D16224.1 | - | 3 - 4 | 315 | 75 | |
U08092.1 | - | 3 - 4 | 75 |