Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF1B
Entrez Gene ID
23095
Gene Name
kinesin family member 1B
Gene Aliases
CMT2, CMT2A, CMT2A1, HMSNII, KLP, NBLST1
Chromosome Location
Chr.1: 10210706 - 10381603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015074.3 NP_055889.2 26 - 27 3158 71
GenBank mRNA AB011163.2 - 28 - 29 3245 71
AB017133.1 - 27 - 28 3128 71
AB088210.1 - 27 - 28 3044 71
AB088211.1 - 26 - 27 3002 71
AB088212.1 - 27 - 28 2966 71
AB088213.1 - 26 - 27 2924 71
AF257176.1 - 26 - 27 3059 71
AK297439.1 - 2 - 3 379 71

View Full Product Details