The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015074.3 | NP_055889.2 | 28 - 29 | 3367 | 81 |
GenBank mRNA | AB011163.2 | - | 30 - 31 | 3454 | 81 |
AB017133.1 | - | 29 - 30 | 3337 | 81 | |
AB088210.1 | - | 29 - 30 | 3253 | 81 | |
AB088211.1 | - | 28 - 29 | 3211 | 81 | |
AB088212.1 | - | 29 - 30 | 3175 | 81 | |
AB088213.1 | - | 28 - 29 | 3133 | 81 | |
AF257176.1 | - | 28 - 29 | 3268 | 81 | |
AK297439.1 | - | 4 - 5 | 588 | 81 |