Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF1B
Entrez Gene ID
23095
Gene Name
kinesin family member 1B
Gene Aliases
CMT2, CMT2A, CMT2A1, HMSNII, KLP, NBLST1
Chromosome Location
Chr.1: 10210706 - 10381603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015074.3 NP_055889.2 33 - 34 3917 61
GenBank mRNA AB011163.2 - 35 - 36 4004 61
AB017133.1 - 34 - 35 3887 61
AB088210.1 - 34 - 35 3803 61
AB088211.1 - 33 - 34 3761 61
AB088212.1 - 34 - 35 3725 61
AB088213.1 - 33 - 34 3683 61
AF257176.1 - 33 - 34 3818 61
AK074279.1 - 3 - 4 257 61
AK297439.1 - 9 - 10 1138 61
BC001415.2 - 1 - 2 93 61
BT007174.1 - 1 - 2 92 61

View Full Product Details