Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBBX
Entrez Gene ID
79740
Gene Name
zinc finger B-box domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 167178399 - 167380405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199201.1 NP_001186130.1 20 - 21 2597 137
NM_001199202.1 NP_001186131.1 19 - 20 2428 137
NM_024687.3 NP_078963.2 19 - 20 2480 137
XM_011513144.1 XP_011511446.1 21 - 22 2642 137
XM_011513145.2 XP_011511447.2 20 - 21 2668 137
XM_011513148.2 XP_011511450.2 21 - 22 2639 137
XM_011513149.2 XP_011511451.2 22 - 23 2759 137
XM_011513151.2 XP_011511453.2 23 - 24 2948 137
XM_011513152.2 XP_011511454.2 15 - 16 2069 137
XM_011513153.2 XP_011511455.2 12 - 13 1741 137
XM_017007186.1 XP_016862675.1 21 - 22 2642 137
XM_017007187.1 XP_016862676.1 20 - 21 2597 137
XM_017007188.1 XP_016862677.1 19 - 20 2480 137
XM_017007189.1 XP_016862678.1 18 - 19 2506 137
GenBank mRNA AK026702.1 - 11 - 12 1446 137
AK093296.1 - 19 - 20 2466 137
BC034229.1 - 20 - 21 2427 137
BC144444.1 - 19 - 20 2544 137

View Full Product Details