The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122752.1 | NP_001116224.1 | 1 - 2 | 179 | 87 |
GenBank mRNA | AK290082.1 | - | 1 - 2 | 102 | 87 |
BC018043.1 | - | 1 - 2 | 100 | 87 | |
DA359846.1 | - | 1 - 2 | 179 | 87 | |
Z81326.1 | - | 1 - 2 | 62 | 87 |