The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122752.1 | NP_001116224.1 | 4 - 5 | 868 | 70 |
NM_005025.4 | NP_005016.1 | 4 - 5 | 1101 | 70 | |
XM_017006618.1 | XP_016862107.1 | 4 - 5 | 1166 | 70 | |
GenBank mRNA | AK290082.1 | - | 4 - 5 | 791 | 70 |
BC018043.1 | - | 4 - 5 | 789 | 70 | |
CR627434.1 | - | 4 - 5 | 1090 | 70 | |
KU178259.1 | - | 3 - 4 | 670 | 70 | |
Z81326.1 | - | 4 - 5 | 751 | 70 |