The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122752.1 | NP_001116224.1 | 7 - 8 | 1266 | 71 |
NM_005025.4 | NP_005016.1 | 7 - 8 | 1499 | 71 | |
XM_017006618.1 | XP_016862107.1 | 7 - 8 | 1564 | 71 | |
GenBank mRNA | AK290082.1 | - | 7 - 8 | 1189 | 71 |
BC018043.1 | - | 7 - 8 | 1187 | 71 | |
CR627434.1 | - | 7 - 8 | 1488 | 71 | |
KU178259.1 | - | 6 - 7 | 1068 | 71 | |
KU178260.1 | - | 3 - 4 | 449 | 71 | |
Z81326.1 | - | 7 - 8 | 1149 | 71 |