The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004911.4 | NP_004902.1 | 7 - 8 | 1412 | 76 |
GenBank mRNA | AK290971.1 | - | 7 - 8 | 1197 | 76 |
AK313299.1 | - | 7 - 8 | 1384 | 76 | |
BC000425.2 | - | 7 - 8 | 1174 | 76 | |
BC001928.1 | - | 7 - 8 | 1174 | 76 | |
BC006344.2 | - | 7 - 8 | 1362 | 76 | |
BC011754.2 | - | 7 - 8 | 1363 | 76 |