Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN7A
Entrez Gene ID
6332
Gene Name
sodium voltage-gated channel alpha subunit 7
Gene Aliases
NaG, Nav2.1, Nav2.2, SCN6A
Chromosome Location
Chr.2: 166403573 - 166494264 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002976.3 NP_002967.2 9 - 10 1214 149
NR_045628.1 9 - 10 1214 149
XM_006712680.2 XP_006712743.1 9 - 10 1283 149
XM_006712681.3 XP_006712744.1 10 - 11 1508 149
XM_006712682.3 XP_006712745.1 11 - 12 1633 149
XM_011511615.2 XP_011509917.1 11 - 12 1579 149
XM_017004667.1 XP_016860156.1 11 - 12 1552 149
GenBank mRNA AK308450.1 - 9 - 10 1205 149
BC062699.1 - 9 - 10 1340 149
BC141864.1 - 9 - 10 1216 149

View Full Product Details