Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHACTR1
Entrez Gene ID
221692
Gene Name
phosphatase and actin regulator 1
Gene Aliases
RPEL, RPEL1, dJ257A7.2
Chromosome Location
Chr.6: 12716635 - 13290484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242648.2 NP_001229577.1 10 - 11 1724 65
NM_001322308.1 NP_001309237.1 9 - 10 1679 65
NM_001322309.1 NP_001309238.1 9 - 10 1570 65
NM_001322310.1 NP_001309239.1 9 - 10 1645 65
NM_001322311.1 NP_001309240.1 7 - 8 1331 65
NM_001322312.1 NP_001309241.1 7 - 8 1694 65
NM_001322313.1 NP_001309242.1 8 - 9 1901 65
NM_001322314.1 NP_001309243.1 8 - 9 1833 65
NM_030948.3 NP_112210.1 10 - 11 1914 65
XM_005248934.2 XP_005248991.1 11 - 12 1963 65
XM_017010452.1 XP_016865941.1 8 - 9 1627 65
XM_017010454.1 XP_016865943.1 11 - 12 1963 65
XM_017010455.1 XP_016865944.1 11 - 12 1931 65
XM_017010456.1 XP_016865945.1 10 - 11 1928 65
XM_017010457.1 XP_016865946.1 10 - 11 1907 65
XM_017010458.1 XP_016865947.1 7 - 8 1420 65
XM_017010459.1 XP_016865948.1 10 - 11 1756 65
XM_017010460.1 XP_016865949.1 10 - 11 1724 65
XM_017010461.1 XP_016865950.1 9 - 10 1695 65
XM_017010462.1 XP_016865951.1 9 - 10 1721 65
XM_017010463.1 XP_016865952.1 8 - 9 1821 65
XM_017010464.1 XP_016865953.1 7 - 8 1420 65
XM_017010465.1 XP_016865954.1 8 - 9 1601 65
XM_017010466.1 XP_016865955.1 8 - 9 1614 65
XM_017010467.1 XP_016865956.1 7 - 8 1601 65
XM_017010469.1 XP_016865958.1 7 - 8 1403 65
GenBank mRNA AB051520.1 - 9 - 10 1521 65
AK090769.1 - 9 - 10 1581 65
AK290017.1 - 10 - 11 1575 65
BC101526.1 - 9 - 10 1488 65
BC101528.1 - 9 - 10 1488 65

View Full Product Details