The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000135.2 | NP_000126.2 | 30 - 31 | 3028 | 77 |
NM_001286167.1 | NP_001273096.1 | 30 - 31 | 3028 | 77 | |
XM_005256294.4 | XP_005256351.1 | 30 - 31 | 3029 | 77 | |
XM_011522946.2 | XP_011521248.1 | 30 - 31 | 3036 | 77 | |
XM_011522947.2 | XP_011521249.1 | 24 - 25 | 2947 | 77 | |
GenBank mRNA | AK299282.1 | - | 30 - 31 | 3017 | 77 |
X99226.1 | - | 30 - 31 | 3017 | 77 |