Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS9D1
Entrez Gene ID
9605
Gene Name
VPS9 domain containing 1
Gene Aliases
ATP-BL, C16orf7
Chromosome Location
Chr.16: 89707133 - 89721046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004913.2 NP_004904.2 12 - 13 1723 121
XM_005256329.4 XP_005256386.1 12 - 13 1786 121
XM_005256330.4 XP_005256387.1 12 - 13 1603 121
XM_005256331.2 XP_005256388.1 9 - 10 1260 121
XM_006721350.2 XP_006721413.1 12 - 13 1786 121
XM_011523476.2 XP_011521778.1 12 - 13 1783 121
XM_011523477.2 XP_011521779.1 11 - 12 3599 121
XM_011523478.2 XP_011521780.1 11 - 12 3596 121
XM_011523480.2 XP_011521782.1 12 - 13 1640 121
GenBank mRNA AB018551.1 - 11 - 12 1762 121
BM920493.1 - 3 - 4 588 121

View Full Product Details