The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004913.2 | NP_004904.2 | 13 - 14 | 1826 | 125 |
XM_005256329.4 | XP_005256386.1 | 13 - 14 | 1889 | 125 | |
XM_005256330.4 | XP_005256387.1 | 13 - 14 | 1706 | 125 | |
XM_005256331.2 | XP_005256388.1 | 10 - 11 | 1363 | 125 | |
XM_011523476.2 | XP_011521778.1 | 13 - 14 | 1886 | 125 | |
XM_011523477.2 | XP_011521779.1 | 12 - 13 | 3702 | 125 | |
XM_011523478.2 | XP_011521780.1 | 12 - 13 | 3699 | 125 | |
XM_011523480.2 | XP_011521782.1 | 13 - 14 | 1743 | 125 | |
GenBank mRNA | AB018551.1 | - | 12 - 13 | 1864 | 124 |
BM920493.1 | - | 4 - 5 | 691 | 125 |