Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS9D1
Entrez Gene ID
9605
Gene Name
VPS9 domain containing 1
Gene Aliases
ATP-BL, C16orf7
Chromosome Location
Chr.16: 89707133 - 89721046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004913.2 NP_004904.2 8 - 9 875 62
XM_005256329.4 XP_005256386.1 8 - 9 935 62
XM_005256330.4 XP_005256387.1 8 - 9 752 62
XM_005256331.2 XP_005256388.1 5 - 6 409 62
XM_006721350.2 XP_006721413.1 8 - 9 935 62
XM_011523476.2 XP_011521778.1 8 - 9 932 62
XM_011523477.2 XP_011521779.1 7 - 8 2748 62
XM_011523478.2 XP_011521780.1 7 - 8 2745 62
XM_011523479.2 XP_011521781.1 8 - 9 935 62
XM_011523480.2 XP_011521782.1 8 - 9 789 62
GenBank mRNA AB018551.1 - 7 - 8 916 62
BY795348.2 - 5 - 6 496 62

View Full Product Details