Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS9D1
Entrez Gene ID
9605
Gene Name
VPS9 domain containing 1
Gene Aliases
ATP-BL, C16orf7
Chromosome Location
Chr.16: 89707133 - 89721046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004913.2 NP_004904.2 10 - 11 1388 80
GenBank mRNA BM920493.1 - 1 - 2 253 80

View Full Product Details