The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013839.3 | NP_001013861.1 | 14 - 15 | 1708 | 72 |
NM_001145297.3 | NP_001138769.1 | 15 - 16 | 1861 | 72 | |
NM_001145298.3 | NP_001138770.1 | 14 - 15 | 1684 | 72 | |
NM_001145299.3 | NP_001138771.1 | 15 - 16 | 1777 | 72 | |
NM_001282313.1 | NP_001269242.1 | 14 - 15 | 1737 | 72 | |
NM_015219.4 | NP_056034.2 | 13 - 14 | 1615 | 72 | |
XM_006721786.3 | XP_006721849.1 | 16 - 17 | 1815 | 72 | |
XM_006721787.3 | XP_006721850.1 | 15 - 16 | 2181 | 72 | |
XM_006721788.3 | XP_006721851.1 | 15 - 16 | 1722 | 72 | |
XM_006721789.3 | XP_006721852.1 | 14 - 15 | 1653 | 72 | |
XM_011524571.2 | XP_011522873.1 | 15 - 16 | 2092 | 72 | |
XM_017024397.1 | XP_016879886.1 | 11 - 12 | 1217 | 72 | |
GenBank mRNA | AB028990.1 | - | 14 - 15 | 1631 | 72 |
AK022552.1 | - | 5 - 6 | 399 | 72 | |
AK023832.1 | - | 13 - 14 | 1523 | 72 | |
AK098284.1 | - | 5 - 6 | 1880 | 72 | |
AK295871.1 | - | 14 - 15 | 1689 | 72 | |
AL834324.1 | - | 14 - 15 | 1679 | 72 | |
AY869728.1 | - | 11 - 12 | 1411 | 72 | |
BC011045.1 | - | 14 - 15 | 1642 | 72 | |
BC018466.1 | - | 15 - 16 | 1769 | 72 | |
BX647763.1 | - | 12 - 13 | 1376 | 72 | |
FJ457119.1 | - | 14 - 15 | 1589 | 72 | |
FJ457120.1 | - | 15 - 16 | 1682 | 72 |