The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290263.1 | NP_001277192.1 | 12 - 13 | 1576 | 96 |
NM_014689.2 | NP_055504.2 | 12 - 13 | 1719 | 96 | |
XM_005246673.4 | XP_005246730.1 | 12 - 13 | 1578 | 96 | |
XM_006712617.3 | XP_006712680.1 | 12 - 13 | 1743 | 96 | |
XM_017004441.1 | XP_016859930.1 | 12 - 13 | 1746 | 96 | |
XM_017004442.1 | XP_016859931.1 | 12 - 13 | 1749 | 96 | |
XM_017004443.1 | XP_016859932.1 | 12 - 13 | 1795 | 96 | |
XM_017004444.1 | XP_016859933.1 | 12 - 13 | 1747 | 96 | |
XM_017004445.1 | XP_016859934.1 | 12 - 13 | 1576 | 96 | |
XM_017004446.1 | XP_016859935.1 | 12 - 13 | 1747 | 96 | |
XM_017004447.1 | XP_016859936.1 | 12 - 13 | 1745 | 96 | |
GenBank mRNA | AB014594.2 | - | 12 - 13 | 1719 | 96 |
EU236710.2 | - | 12 - 13 | 1573 | 96 |