The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290263.1 | NP_001277192.1 | 36 - 37 | 4144 | 81 |
NM_014689.2 | NP_055504.2 | 36 - 37 | 4287 | 81 | |
XM_005246673.4 | XP_005246730.1 | 36 - 37 | 4146 | 81 | |
XM_006712617.3 | XP_006712680.1 | 36 - 37 | 4311 | 81 | |
XM_017004441.1 | XP_016859930.1 | 36 - 37 | 4314 | 81 | |
XM_017004442.1 | XP_016859931.1 | 36 - 37 | 4290 | 81 | |
XM_017004443.1 | XP_016859932.1 | 36 - 37 | 4363 | 81 | |
XM_017004444.1 | XP_016859933.1 | 35 - 36 | 4270 | 81 | |
XM_017004445.1 | XP_016859934.1 | 36 - 37 | 4144 | 81 | |
XM_017004446.1 | XP_016859935.1 | 35 - 36 | 4168 | 81 | |
XM_017004447.1 | XP_016859936.1 | 36 - 37 | 4286 | 81 | |
GenBank mRNA | AK293847.1 | - | 2 - 3 | 213 | 81 |
AK293879.1 | - | 7 - 8 | 693 | 81 | |
CR749492.1 | - | 11 - 12 | 1132 | 81 | |
EU236710.2 | - | 36 - 37 | 4141 | 81 |