Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBBX
Entrez Gene ID
79740
Gene Name
zinc finger B-box domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 167178399 - 167380405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199202.1 NP_001186131.1 3 - 4 231 147
XM_011513149.2 XP_011511451.2 4 - 5 400 147
XM_011513151.2 XP_011511453.2 4 - 5 444 147
GenBank mRNA BC034229.1 - 3 - 4 231 147

View Full Product Details