Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBBX
Entrez Gene ID
79740
Gene Name
zinc finger B-box domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 167178399 - 167380405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199201.1 NP_001186130.1 5 - 6 516 81
NM_001199202.1 NP_001186131.1 5 - 6 464 81
NM_024687.3 NP_078963.2 5 - 6 516 81
XM_011513144.1 XP_011511446.1 5 - 6 516 81
XM_011513145.2 XP_011511447.2 4 - 5 542 81
XM_011513148.2 XP_011511450.2 5 - 6 516 81
XM_011513149.2 XP_011511451.2 6 - 7 633 81
XM_011513155.1 XP_011511457.1 5 - 6 516 81
XM_011513156.1 XP_011511458.1 5 - 6 516 81
XM_017007186.1 XP_016862675.1 5 - 6 516 81
XM_017007187.1 XP_016862676.1 5 - 6 516 81
XM_017007188.1 XP_016862677.1 5 - 6 516 81
XM_017007189.1 XP_016862678.1 4 - 5 542 81
XM_017007190.1 XP_016862679.1 5 - 6 516 81
GenBank mRNA AK093296.1 - 5 - 6 502 81
BC034229.1 - 5 - 6 464 81
BC144444.1 - 4 - 5 463 81
DB078376.1 - 5 - 6 516 81

View Full Product Details