Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 21 - 22 3702 132
NM_001322988.1 NP_001309917.1 21 - 22 3702 132
NM_001322989.1 NP_001309918.1 21 - 22 3675 132
NM_001322990.1 NP_001309919.1 21 - 22 3663 132
NM_001322991.1 NP_001309920.1 21 - 22 3636 132
NM_001322992.1 NP_001309921.1 20 - 21 3552 132
NM_003947.5 NP_003938.1 21 - 22 3702 132
XM_006713810.3 XP_006713873.1 21 - 22 3947 132
XM_006713811.3 XP_006713874.1 21 - 22 3920 132
XM_006713812.3 XP_006713875.1 21 - 22 3908 132
XM_006713813.3 XP_006713876.1 21 - 22 3881 132
XM_006713814.3 XP_006713877.1 21 - 22 3947 132
XM_006713815.3 XP_006713878.1 21 - 22 3950 132
XM_011513279.2 XP_011511581.1 21 - 22 3947 132
XM_011513280.2 XP_011511582.1 21 - 22 3948 132
XM_011513281.2 XP_011511583.1 21 - 22 26569 132
XM_011513283.2 XP_011511585.1 21 - 22 3944 132
XM_011513285.2 XP_011511587.1 21 - 22 3943 132
XM_017007429.1 XP_016862918.1 21 - 22 3947 132
XM_017007430.1 XP_016862919.1 21 - 22 3947 132
XM_017007431.1 XP_016862920.1 21 - 22 3943 132
XM_017007432.1 XP_016862921.1 21 - 22 3943 132
XM_017007433.1 XP_016862922.1 21 - 22 3923 132
XM_017007434.1 XP_016862923.1 21 - 22 3911 132
XM_017007435.1 XP_016862924.1 21 - 22 3884 132
GenBank mRNA AK131379.1 - 11 - 12 1763 132
AK314126.1 - 21 - 22 3675 132
U94190.1 - 21 - 22 3645 132

View Full Product Details