Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 22 - 23 3885 88
NM_001322988.1 NP_001309917.1 22 - 23 3885 88
NM_001322989.1 NP_001309918.1 22 - 23 3858 88
NM_001322990.1 NP_001309919.1 22 - 23 3846 88
NM_001322991.1 NP_001309920.1 22 - 23 3819 88
NM_001322992.1 NP_001309921.1 21 - 22 3735 88
NM_003947.5 NP_003938.1 22 - 23 3885 88
XM_006713810.3 XP_006713873.1 22 - 23 4130 88
XM_006713811.3 XP_006713874.1 22 - 23 4103 88
XM_006713812.3 XP_006713875.1 22 - 23 4091 88
XM_006713813.3 XP_006713876.1 22 - 23 4064 88
XM_006713814.3 XP_006713877.1 22 - 23 4130 88
XM_006713815.3 XP_006713878.1 22 - 23 4133 88
XM_011513279.2 XP_011511581.1 22 - 23 4130 88
XM_011513280.2 XP_011511582.1 22 - 23 4131 88
XM_011513281.2 XP_011511583.1 22 - 23 26752 88
XM_011513283.2 XP_011511585.1 22 - 23 4127 88
XM_011513285.2 XP_011511587.1 22 - 23 4126 88
XM_017007429.1 XP_016862918.1 22 - 23 4130 88
XM_017007430.1 XP_016862919.1 22 - 23 4130 88
XM_017007431.1 XP_016862920.1 22 - 23 4126 88
XM_017007432.1 XP_016862921.1 22 - 23 4126 88
XM_017007433.1 XP_016862922.1 22 - 23 4106 88
XM_017007434.1 XP_016862923.1 22 - 23 4094 88
XM_017007435.1 XP_016862924.1 22 - 23 4067 88
GenBank mRNA AK131379.1 - 12 - 13 1946 88
AK314126.1 - 22 - 23 3858 88
U94190.1 - 22 - 23 3828 88

View Full Product Details