The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319958.1 | NP_001306887.1 | 2 - 3 | 505 | 127 |
XM_011541713.2 | XP_011540015.1 | 1 - 2 | 3980 | 127 | |
XM_011541715.2 | XP_011540017.1 | 2 - 3 | 428 | 127 | |
XM_011541726.2 | XP_011540028.1 | 1 - 2 | 3980 | 127 | |
XM_017001639.1 | XP_016857128.1 | 2 - 3 | 219 | 127 | |
XM_017001642.1 | XP_016857131.1 | 1 - 2 | 3980 | 127 | |
GenBank mRNA | BC038389.1 | - | 2 - 3 | 423 | 127 |