The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198961.1 | NP_001185890.1 | 4 - 5 | 468 | 109 |
XM_011541709.2 | XP_011540011.1 | 5 - 6 | 679 | 109 | |
XM_011541710.2 | XP_011540012.1 | 5 - 6 | 679 | 109 | |
XM_011541712.2 | XP_011540014.1 | 5 - 6 | 679 | 109 | |
XM_011541713.2 | XP_011540015.1 | 4 - 5 | 4456 | 109 | |
XM_011541715.2 | XP_011540017.1 | 5 - 6 | 904 | 109 | |
XM_011541719.2 | XP_011540021.1 | 5 - 6 | 1901 | 109 | |
XM_011541720.2 | XP_011540022.1 | 4 - 5 | 4225 | 109 | |
XM_011541721.2 | XP_011540023.1 | 6 - 7 | 918 | 109 | |
XM_011541722.2 | XP_011540024.1 | 5 - 6 | 536 | 109 | |
XM_011541723.2 | XP_011540025.1 | 5 - 6 | 636 | 109 | |
XM_017001638.1 | XP_016857127.1 | 6 - 7 | 2290 | 109 | |
XM_017001639.1 | XP_016857128.1 | 5 - 6 | 695 | 109 | |
XM_017001640.1 | XP_016857129.1 | 4 - 5 | 858 | 109 | |
XM_017001641.1 | XP_016857130.1 | 6 - 7 | 2059 | 109 | |
GenBank mRNA | AK299834.1 | - | 5 - 6 | 637 | 109 |
AK300896.1 | - | 5 - 6 | 507 | 109 | |
AK302285.1 | - | 7 - 8 | 1047 | 109 | |
AK315906.1 | - | 5 - 6 | 599 | 109 | |
BC044574.1 | - | 4 - 5 | 109 | ||
BC051887.1 | - | 4 - 5 | 456 | 109 |