The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146029.2 | NP_001139501.1 | 12 - 13 | 1645 | 70 |
NM_001146030.2 | NP_001139502.1 | 13 - 14 | 1583 | 70 | |
NM_003612.4 | NP_003603.1 | 13 - 14 | 1687 | 70 | |
GenBank mRNA | AF030698.1 | - | 13 - 14 | 1658 | 70 |
AF069493.1 | - | 13 - 14 | 1663 | 70 | |
AF071542.1 | - | 13 - 14 | 1646 | 70 | |
AK001895.1 | - | 7 - 8 | 928 | 70 | |
AK293280.1 | - | 12 - 13 | 1641 | 70 | |
AK312257.1 | - | 13 - 14 | 1686 | 70 | |
AM180445.1 | - | 13 - 14 | 1646 | 70 | |
AM180446.1 | - | 13 - 14 | 1646 | 70 | |
AM180447.1 | - | 13 - 14 | 1646 | 70 | |
AM180448.1 | - | 13 - 14 | 1646 | 70 | |
AM180449.1 | - | 13 - 14 | 1646 | 70 | |
AM180450.1 | - | 13 - 14 | 1646 | 70 | |
AM180451.1 | - | 13 - 14 | 1646 | 70 | |
BC101643.1 | - | 13 - 14 | 1678 | 70 | |
BC101647.1 | - | 13 - 14 | 1678 | 70 |